trans-N-(6,6-Dimethyl-2-hepten-4-ynyl)-N-methyl-1-naphthylmethylamine hydrochloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Terbinafine Hydrochloride, Terbinafine HCl, (E)-N,6,6-Trimethyl-N-(naphthalen-1-ylmethyl)hept-2-en-4-yn-1-amine hydrochloride
CAS Number:78628-80-5
:
Formula:C₂₁H₂₅N
MDL Number:MFCD00145430
:327.90 g/mol
Melting Pt:214 °C
Storage Temperature:Ambient
UN:3077
ADR:9,III
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (1 g, 105)
- Solubility:(5 % in methanol) Clear solution
- HPLC:≥98.5 %
Specification Test Results
Frequently Bought Together