1,5-Anhydro-D-sorbitol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1-Deoxy-D-glucose, 1,5-Anhydro-D-glucitol
CAS Number:154-58-5
:
Formula:C₆H₁₂O₅
MDL Number:MFCD00067386
:164.16 g/mol
Melting Pt:141…144 °C
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Powder or crystalline powder
- Infrared spectrum:Conforms
- Melting point:140 to 145 °C
- Elemental Analysis:≥42.14 to ≤45.66 % Carbon
Specification Test Results
Frequently Bought Together