
1-Boc-piperidin-4-ylboronic acid pinacol ester 97%
ACRO456050050
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:White to yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:60 to 65 °C
- HPLC:≥96.0 %
:
:311.2247 g/mol
:56.5…57 °C
:GHS07
:Refrigerator
:1048970-17-7
:C₁₆H₃₀BNO₄
:MFCD09997792