Order Entry
Switzerland
Orders LinkContactUsLinkComponent
6-(Hydroxymethyl)indole 97%

6-(Hydroxymethyl)indole 97%

Irritant
6-(Hydroxymethyl)indole 97%
Certificates
6-(Hydroxymethyl)indole 97%

2 Options Available Below

Product Details & Documents
Synonyms:  (1H-Indol-6-yl)methanol
  • CAS Number:1075-26-9
  • Formula:C₉H₉NO
  • MDL Number:MFCD02179595
  • Molecular Weight:147.1739 g/mol
  • Melting Pt:45…50 °C
  • Storage Temperature:Refrigerator
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    60 to 67 °C
  • Water:
    ≤1 %
  • GC:
    ≥96.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - SFr. 110.00
    SFr. 110.00
    1 * 250 mg
       SFr. 110.00

  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - SFr. 295.00
    SFr. 295.00
    1 * 1 g
       SFr. 295.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.