Order Entry
Switzerland
ContactUsLinkComponent
Tris(hydroxypropyl)phosphine ≥80% (by 31P-NMR), Millipore®
Corrosive
Tris(hydroxypropyl)phosphine ≥80% (by 31P-NMR), Millipore®
Catalog # 598250-1
CAS Number:  
Corrosive
Tris(hydroxypropyl)phosphine ≥80% (by 31P-NMR), Millipore®
Catalog # 598250-1
CAS Number:  

Specifications

  • Pack type:
    Aluminium drum
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Form:
    Colorless viscous liquid
  • Purity:
    ≥80% by P-NMR
  • Storage:
    +2 °C to +8 °C

Specification Test Results

About this item

 :
 :208.24 g/mol
ADR:8,II
 :Packaged under inert gas.
UN:3265
 :GHS05
Storage Temperature:Refrigerator
CAS Number:4706-17-6
Formula:C₉H₂₁O₃P
MDL Number:MFCD00269835