SATA (N-Succinimidyl S-Acetylthioglycolate) ≥98% (durch HPLC), weißer Feststoff, Millipore®
Catalog # 573100-100
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Packung:Plastic ampoule
- VE:100 mg
- Seq:2440
Specifications
Specification Test Results
- Form:White solid
- Purity:≥98% by HPLC
- Storage:–20 °C
- Stability:Stable for up to 3 months at –20 °C
Specification Test Results
About this item
:
:231.22 g/mol
CAS Number:76931-93-6
Formula:C₈H₉NO₅S
MDL Number:MFCD00036891