Coloured slide boxes
Supplier: GLW
Jump to:
About this item
Slide boxes with 25 compartments and snap lock provide a secure hold for the slides and coverslips.
- Stackable for use as a minimum space as possible
- Can be used in a freezer down to –80 °C
- Numbers in the bottom of the box and hinged lid
Designed for storing, transporting, and freezing the samples.
Ordering information: Dividers must be ordered separately.
9 Options Available Below
Product Details & Documents
Slide boxes with 25 compartments and snap lock provide a secure hold for the slides and coverslips.
- Stackable for use as a minimum space as possible
- Can be used in a freezer down to –80 °C
- Numbers in the bottom of the box and hinged lid
Designed for storing, transporting, and freezing the samples.
Ordering information: Dividers must be ordered separately.
Product Family Options
Product Information
- ColourMaterialNo. of slidesW×D×HAvailabilityPrice
- GLWAK25BSlide box with snap lock
- GLWAK25GSlide box with snap lock
- GLWAK25OSlide box with snap lock
- GLWAK25RSlide box with snap lock
- GLWAK25SSlide box with snap lock
- GLWAK25TSlide box with snap lock
- GLWAK25VSlide box with snap lock
- GLWAK25WSlide box with snap lock
- GLWAK25YSlide box with snap lock
Accessories
Product Information
- Catalog #ColourMaterialAvailabilityPrice
- Divider for slides and coverslips of special sizesDivider for slides and coverslips of special sizesGLWAZK25WhitePSLog in for availability1 * 1 ST - SFr. 3.00
Product Information
- Catalog #ColourMaterialAvailabilityPrice
- Divider for slides and coverslips of special sizesDivider for slides and coverslips of special sizesGLWAZK20GreyPSLog in for availability1 * 1 ST - SFr. 3.10