Order Entry
Switzerland
ContactUsLinkComponent
Disposable particle filtering respirators, C-series
Catalog # ALPSC-3V
Anbieter:  Alpha Solway
undefined
Disposable particle filtering respirators, C-series
Catalog # ALPSC-3V
Anbieter:  Alpha Solway

Technische Daten

  • Type:
    C-3V, orange, oil water based particles, very fine toxic dusts/mists/fumes, metal fumes
  • Seq:
    02
  • Pk:
    1
  • With valve:
    +
  • Class:
    FFP3

Technische Daten

Über diesen Artikel

This FFP2 mask is a lightweight low profile twin fold disposable respirator featuring a unique pleat design that enables the wearer to talk without compromising fit. The half-mask is made from a PP Non-Woven inner and outer layer, a polyester central layer and a high filtration efficiency melt-blown PP filter media.

  • Twin fold design, flexes whilst talking to maintain seal and enables easy storage in a pocket
  • Soft lightweight fabric provides comfortable face seal
  • Large surface area of filter media lowers breathing resistance
  • Low profile exhalation valve to reduce heat and moisture build up
  • Colour coded filter media for ease of identification

Protects from oil or water based particulates, toxic dusts (and metal fumes). Ideal fo use in hospitals, laboratory and disease control or iron and steel industry.

Certifications: EN 149:2001 + A1:2009 FFP2 NR D