Standards for water pollution QC check, hexavalent chromium, VHG
Catalog # PROOVHG-QWPCR6-15
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:15 mL
- Description:Water analysis standard for hexavalent chromium
- Type:Reference standard
Specifications
About this item
This solution is intended for use as a quality control check for methods related to hexavalent chromium.
This standard has been prepared as a concentrate and must be diluted prior to analysis and should be analysed as soon as possible after the concentrate is diluted. This concentrate is not preserved but can be stored at room temperature.
Certifications: Traceable to NIST SRM.