Order Entry
Switzerland
ContactUsLinkComponent
Sulphide standard solution, for IC, CRM
Sulphide standard solution, for IC, CRM
Catalog # PROORTCICS023-100
CAS Number:  
Sulphide standard solution, for IC, CRM
Catalog # PROORTCICS023-100
CAS Number:  

Specifications

  • Concentration:
    1000 mg/l
  • Matrix:
    NaOH 1%
  • Pk:
    100 mL
  • Description:
    Sulphide standard solution

Specifications

About this item

Certifications: Traceable to NIST, SRM.