ContactUsLinkComponent
Vacuum adapter, GL thread, PTFE, BOLA
Vacuum adapter, GL thread, PTFE, BOLA
Catalog # BOHLN1656-45
Supplier:  Bohlender
undefined
Vacuum adapter, GL thread, PTFE, BOLA
Catalog # BOHLN1656-45
Supplier:  Bohlender
Specifications
  • Hole Ø:
    22 mm
  • Connector:
    GL 45
  • Pk:
    1
Specifications
About this item
This vacuum adapter is made of PTFE with O-ring made of FKM for secure placement of standard filter cartridges made of glass or plastic on flasks with GL 45 thread.
  • Lateral step-type solenoid made of PTFE Chemical resistance Temperature resistance from -15 to +200 °C
For vacuum filtration, the O-ring at the passageway seals the connection to the filter slide so that even a small vacuum is sufficient.