ContactUsLinkComponent
Elemental analysis standards, PTP sample of oils, VHG
Elemental analysis standards, PTP sample of oils, VHG
Catalog # PROOVHG-VPTPMO-25G
CAS Number:  
Elemental analysis standards, PTP sample of oils, VHG
Catalog # PROOVHG-VPTPMO-25G
CAS Number:  
Specifications
  • Pk:
    25 g
  • Description:
    PTP sample of oils, VHG, 25 g
Specifications
About this item
The PTP has been specially designed for analytical laboratories that analyse new and used oils for metals, sulphur, viscosity and particle count.
  • Monitor and improve measurements processes
  • Demonstrate analytical competency
  • Method and instrumentation validation
  • 25 analytes
Certifications: ISO/IEC 17043 accredited.