Elemental analysis standards, soot content standards, VHG
Supplier: LGC Standards PROMOCHEM
Certificates
About this item
This reference or calibration standard is intended for use in the determination of soot in diesel motor oil by infrared spectroscopy and other technique.
Certifications: Conforms to ISO 9001 and ISO/IEC 17025.
Frequently Bought Together