Barrels with lid and clamping ring
Anbieter: RIXIUS
Zertifikate
Über diesen Artikel
HDPE, blue.
- 30 and 60 litre versions include handles
- Good chemical and temperature resistance
- All sizes with UN approval for solids
Heavy duty open top drums with PE lid and galvanised clamping ring. Suitable for storage, disposal and transport of laboratory materials.
Frequently Bought Together