Pipette tips, Sapphire
Catalog # GREI771255_960
Supplier: Greiner Bio-One
Specifications
- Packed:Rack (96 tips/rack)
- Sterile:-
- Volume:10 µl
- Seq:02
- Tip style:Graduated, low retention
- Tip colour:Transparent
- Pk:960
Specifications
About this item
These tips are manufactured from virgin materials. All tips are transparent, graduated, and allow precise and comfortable pipetting with maximal recovery.
- Graduations for perfect visual control of liquid
- Thin-walled top for optimal seal
- Free of detectable DNase, RNase, human DNA and non pyrogenic
- Available as low retention tips for even higher recovery