Methyl palmitate 99% analytical standard for GC
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112-39-0
:
Formula:C₁₇H₃₄O₂
MDL Number:MFCD00008994
:270.46 g/mol
Boiling Pt:338 °C (760 mmHg)
Melting Pt:28 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Low melting solid
- Infrared spectrum:Conforms
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together