Methanol, J.T.Baker®
: Avantor
:67-56-1
:
:CH₃OH
:MFCD00004595
:32.04 g/mol
:1230
:3,II
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Assay (by GC) (corrected for water):min. 99.8 %
- Appearance:clear
- Carbonyl Compounds (max. 0.001% each of Acetone, Formaldehyde and Acetaldehyde):passes test
- Color (APHA):max. 10
- Residue after Evaporation:max. 0.001 %
- Solubility in Water:passes test
- Substances Darkened by H2SO4:passes test
- Substances Reducing KMnO4:passes test
- Titrable Acid (meq/g):max. 0.0003
- Titrable Base (meq/g):max. 0.0002
- Water (H2O):max. 0.01 %
- Aluminium (Al):max. 0.5
- Boron (B):max. 0.02
- Barium (Ba):max. 0.1
- Calcium (Ca):max. 0.5
- Cadmium (Cd):max. 0.05
- Cobalt (Co):max. 0.02
- Chromium (Cr):max. 0.02
- Copper (Cu):max. 0.02
- Iron (Fe):max. 0.1
- Magnesium (Mg):max. 0.1
- Manganese (Mn):max. 0.02
- Nickel (Ni):max. 0.02
- Lead (Pb):max. 0.1
- Tin (Sn):max. 0.1
- Zinc (Zn):max. 0.1
Frequently Bought Together