Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Alcohol, Reference Standards

Alcohol, Reference Standards

Supplier:   USP
Alcohol, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1012699.
  • 1012688.
  • USPH1012768
  • USPH1012772
  • Description:
  • Alcohol Determination--Acetonitrile
  • Alcohol Determination - Alcohol
  • Alcohol
  • Dehydrated Alcohol
  • CAS No.:
  • 75-05-8
  • 64-17-5
  • 64-17-5
  • 64-17-5
  • Formula:
  • H₃CCN
  • C₂H₆O
  • C₂H₆O
  • C₂H₆O
  • UN No.:
  • 1993
  • 1170
  • 1170
  • 1170

Specifications

Product Family Options

Product Information

  • Standards
    Pk Info
    Pk
    Availability
    Price
  • 1012699.
    Alcohol Determination--Acetonitrile
    Alcohol Determination--Acetonitrile
    USP reference standard
    5 x 5 mL
    5
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 534.00
    SFr. 534.00
    1 * 5 ST
       SFr. 534.00

    Specifications:
    More
    Specifications
    Cat. No.
    1012699.
    Description
    Alcohol Determination--Acetonitrile
    CAS No.
    75-05-8
    Formula
    H₃CCN
    UN No.
    1993
  • 1012688.
    Alcohol Determination - Alcohol
    Alcohol Determination - Alcohol
    USP reference standard
    5 mL/ampule; 5 ampules
    5
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - SFr. 509.00
    SFr. 509.00
    1 * 5 ST
       SFr. 509.00

    Specifications:
    More
    Specifications
    Cat. No.
    1012688.
    Description
    Alcohol Determination - Alcohol
    CAS No.
    64-17-5
    Formula
    C₂H₆O
    UN No.
    1170
  • Alcohol
    USP reference standard
    1.2 mL/ampule; 5 ampules
    5
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    USPH1012768
    Description
    Alcohol
    CAS No.
    64-17-5
    Formula
    C₂H₆O
    UN No.
    1170
  • USPH1012772
    Dehydrated Alcohol
    Dehydrated Alcohol
    USP reference standard
    1.2 mL/ampule; 5 ampules
    5
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    USPH1012772
    Description
    Dehydrated Alcohol
    CAS No.
    64-17-5
    Formula
    C₂H₆O
    UN No.
    1170