Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Optical plate

Optical plate

Optical BK7 plate, 80×50 mm, 15 mm thick
Codice catalogo DIDAPOD010240
Nome del fornitore:   DIDALAB
Optical plate
Codice catalogo DIDAPOD010240
Nome del fornitore:  DIDALAB
Optical plate
Optical plate

Optical BK7 plate, 80×50 mm, 15 mm thick

Price
Richiedi prezzo
Log in per disponibilità
1 * 1 ST
Product Details & Documents

Optical glass components are manufactured with high accuracy polishing and flatness tolerance.

  • Optical polishing
  • Two sides with parallelism less than 1 µm
Specifiche
  • Description:
    Optical BK7 plate, 80×50 mm, 15 mm thick
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.