Test sieves, 100×40 mm
Supplier: ENDECOTTS
New Product
Certificates
About this item
Stainless steel woven wire mesh test sieves are ideal for all types of laboratory sampling and particle size analysis.
For more size options please contact your local Avantor office.
Certifications: ISO3310-1:2000, BS410-1:2000
Delivery: Each sieve is supplied together with a certificate of examination and carries the BS 410 kite mark.
Frequently Bought Together