Order Entry
Switzerland
ContactUsLinkComponent
Standards for water analysis, total kjeldahl nitrogen standard, VHG
Standards for water analysis, total kjeldahl nitrogen standard, VHG
Catalog # PROOG-TOTKJN1K-500
CAS Number:  
Standards for water analysis, total kjeldahl nitrogen standard, VHG
Catalog # PROOG-TOTKJN1K-500
CAS Number:  

Specifications

  • Concentration:
    1000 mg/l in 1% HCl
  • Pk:
    1
  • Description:
    Total kjeldahl nitrogen standard (TKN)

Specifications

About this item

This standard solution is intended for use as a calibration standard for total kjeldahl nitrogen (TKN).

Certifications: Traceable to NIST and SRM.