ContactUsLinkComponent
Gasoline fuel dilution standards, VHG
Catalog # PROOVHGGASFDBLK100
CAS Number:  
Gasoline fuel dilution standards, VHG
Catalog # PROOVHGGASFDBLK100
CAS Number:  
Specifications
  • Pk:
    100 mL
  • Description:
    Blank for as fuel dilution standards in 75 cSt hydrocarbon oil
Specifications
About this item
This standard is intended for use in the determination of gasoline in used gasoline engine oil by gas chromatography (GC), IR, fuel dilution meter (fuel sniffer) and flash point analyser.
It is prepared gravimetrically to contain 0% (v/v) of devolatilised (10% light volatiles removed) gasoline in 75 cSt mineral oil. The certified concentration is based upon the gravimetric preparation of the standard.
Certifications: Accordance with ASTM D3525 and D7593. ISO 9001 and ISO/IEC 17025 accredited. Traceable to NIST.