ContactUsLinkComponent
Electrode stand with swivel arm, Eutech
Electrode stand with swivel arm, Eutech
Katalognr. EUTEECPHELSTDC
Supplier:  EUTECH
undefined
Electrode stand with swivel arm, Eutech
Katalognr. EUTEECPHELSTDC
Supplier:  EUTECH
Specifications
  • For:
    Up to four standard size electrodes and one ATC probe
  • Description:
    Electrode stand with swivel arm
  • Pk:
    1
Specifications
About this item
This electrode stand with swivel arm allows convenient hands-free operation. Holds up to four standard size electrodes and one ATC probe.
  • Weighted 6-inch diameter base allows users to pivot holder 360° for more flexibility
  • Arm allows users to move electrode side to side or up and down while keeping electrode at a constant vertical angle