Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Etoposide, Reference Standards

Etoposide, Reference Standards

Supplier:   USP
Etoposide, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1268808.
  • 1268852.
  • 1268819.
  • Description:
  • Etoposide
  • Etoposide Resolution Mixture
  • Etoposide Phosphate
  • CAS No.:
  • 33419-42-0
  • 33419-42-0
  • 117091-64-2
  • Formula:
  • C₂₉H₃₂O₁₃
  • UN No.:
  • 1325

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1268808.
    Etoposide
    Etoposide
    USP reference standard
    Vial
    300 mg
    Log in for availability
    Log in for availability
    1 * 300 mg
    1 * 300 mg - SFr. 415.00
    SFr. 415.00
    1 * 300 mg
       SFr. 415.00

    Specifications:
    More
    Specifications
    Cat. No.
    1268808.
    Description
    Etoposide
    CAS No.
    33419-42-0
    Formula
    C₂₉H₃₂O₁₃
    UN No.
  • 1268852.
    Etoposide Resolution Mixture
    Etoposide Resolution Mixture
    USP reference standard
    Vial
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - SFr. 1’490.00
    SFr. 1’490.00
    1 * 10 mg
       SFr. 1’490.00

    Specifications:
    More
    Specifications
    Cat. No.
    1268852.
    Description
    Etoposide Resolution Mixture
    CAS No.
    33419-42-0
    Formula
    UN No.
  • 1268819.
    Etoposide Phosphate
    Etoposide Phosphate
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - SFr. 633.00
    SFr. 633.00
    1 * 200 mg
       SFr. 633.00

    Specifications:
    More
    Specifications
    Cat. No.
    1268819.
    Description
    Etoposide Phosphate
    CAS No.
    117091-64-2
    Formula
    UN No.
    1325
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.