Erythromycin, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
4 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1242010.
- 1242021.
- 1242032.
- 1242000.
- Description:
- Erythromycin B
- Erythromycin C
- Erythromycin Related Compound N
- Erythromycin
- Synonym(s):
- N-Demethylerythromycin A
- CAS No.:
- 527-75-3
- 1675-02-1
- 992-62-1
- 114-07-8
- Formula:
- C₃₇H₆₇NO₁₂
- C₃₆H₆₅NO₁₃
- C₃₆H₆₅NO₁₃
- UN No.:
- 3077
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1242010.Erythromycin BSpecifications:
More
Specifications
Cat. No.1242010.DescriptionErythromycin BSynonym(s)CAS No.527-75-3FormulaC₃₇H₆₇NO₁₂UN No. - 1242021.Erythromycin CSpecifications:
More
Specifications
Cat. No.1242021.DescriptionErythromycin CSynonym(s)CAS No.1675-02-1FormulaC₃₆H₆₅NO₁₃UN No. - 1242032.Erythromycin Related Compound NErythromycin Related Compound NUSP reference standardVial50 mgLog in for availability1 * 50 mgSFr. 1’460.00Specifications:
More
Specifications
Cat. No.1242032.DescriptionErythromycin Related Compound NSynonym(s)N-Demethylerythromycin ACAS No.992-62-1FormulaC₃₆H₆₅NO₁₃UN No. - 1242000.ErythromycinSpecifications:
More
Specifications
Cat. No.1242000.DescriptionErythromycinSynonym(s)CAS No.114-07-8FormulaUN No.3077
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



