Order Entry
Switzerland
ContactUsLinkComponent
Methanol, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Methanol, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Methanol, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Assay (by GC) (corrected for water):
    min. 99.8 %
  • Acetone:
    max. 0.001 %
  • Color (APHA):
    max. 10
  • Residue after Evaporation:
    max. 1 ppm
  • Substances Darkened by H2SO4:
    passes test
  • Titrable Acid (meq/g):
    max. 0.0003
  • Titrable Base (meq/g):
    max. 0.0001
  • Water (H2O):
    max. 0.08 %
  • Single Impurity Peak (pg/ml):
    max. 10
  • Single Impurity Peak (ng/ml):
    max. 5

Specification Test Results

Frequently Bought Together