Order Entry
Switzerland
ContactUsLinkComponent
N,N-Dimethylacetamide, anhydrous (max. 0.005% H₂O)
Health
Irritant
N,N-Dimethylacetamide, anhydrous (max. 0.005% H₂O)
Catalog # 85817.230
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Irritant
N,N-Dimethylacetamide, anhydrous (max. 0.005% H₂O)
Catalog # 85817.230
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle with septum cap
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

Specification Test Results

  • Assay (GC, on anhydrous basis):
    99.8-100 %
  • Acidity 0-0.0005:
    0-0.001 %
  • Residue after evaporation:
    0-0.001 % w/w
  • Water (KF):
    0-0.005 % w/w

Specification Test Results

About this item

 :
 :87.12 g/mol
Merck Index:13,03254
Boiling Pt:166.1 °C (1013 hPa)
Melting Pt:–20 °C
Density:0.937 g/cm³ (20 °C)
Flash Pt:70 °C
Storage Temperature:Ambient
EINECS:204-826-4
UN:2810
ADR:6.1,III
REACH:01-2119459339-27
 :GHS08
 :GHS07
CAS Number:127-19-5
Formula:CH₃CON(CH₃)₂
MDL Number:MFCD00008686