

Chloroform, anhydrous (max. 0.005% H₂O) stabilised
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:67-66-3
:
Formula:CHCl₃
MDL Number:MFCD00000826
:119.38 g/mol
Merck Index:13,02160
Boiling Pt:61.7 °C (1013 hPa)
Melting Pt:–63 °C
Density:1.4832 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1888
ADR:6.1,III
:Stabilised with 2-methyl-2-butene (iso-amylene) 0.005 % max.
EINECS:200-663-8
REACH:01-2119486657-20
:GHS08
:GHS06
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Assay (GC, on anhydrous basis):99.9-100 %
- Appearance:Clear colorless liquid
- Acidity (as HCl):0-0.001 %
- Residue after evaporation:0-0.0005 % w/w
- Stabilizer (Amylene):0.003-0.006 % w/w
- Water (KF):0-0.005 % w/w
Specification Test Results
Frequently Bought Together