Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standard, Oxymetazoline hydrochloride, Ph.Eur., RS
Organic reference standard, Oxymetazoline hydrochloride, Ph.Eur., RS
Catalog # EDQMO0290000
Supplier:  EDQM
CAS Number:  
Organic reference standard, Oxymetazoline hydrochloride, Ph.Eur., RS
Catalog # EDQMO0290000
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    80 mg
  • Standards:
    Reference standard
  • CAS No.:
    2315-02-8
  • Description:
    Oxymetazoline hydrochloride
  • Formula:
    C₁₆H₂₅ClN₂O
  • Storage temperature:
    Ambient
  • Description:
    Oxymetazoline hydrochloride

Specifications

About this item

  • In Monograph 0943
  • Solubility in water: 100 to 1000 g/l