Organic reference standard, Methylnitrosoindoline, Ph.Eur., RS
EDQMM1625000
: EDQM
:
- Pack type:Vial
- Pk:110 mg
- Standards:Reference standard
- CAS No.:85440-79-5
- Description:Methylnitrosoindoline
- Formula:C₉H₁₀N₂O
- Storage temperature:Ambient
- Description:Methylnitrosoindoline
- In Monograph 1108
- Solubility in water: less than 1 g/l