Organic reference standard, Ergocalciferol, Ph.Eur., RS
Catalog # EDQME0900000
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Vial
- Pk:150 mg
- Standards:Reference standard
- CAS No.:50-14-6
- Description:Ergocalciferol
- Formula:C₂₈H₄₄O
- Storage temperature:-20°C ± 5°C
- Description:Ergocalciferol
Specifications
About this item
Filled under inert atmosphere protected from humidity.
- In Monographs 0082, 0574, 0575, 0598, 1192, 1193, and 2398