Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standards, Dextromethorphan, Ph.Eur., RS
Organic reference standards, Dextromethorphan, Ph.Eur., RS
Catalog # EDQMY0000261
Supplier:  EDQM
CAS Number:  
Organic reference standards, Dextromethorphan, Ph.Eur., RS
Catalog # EDQMY0000261
Supplier:  EDQM
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    5 mg
  • Standards:
    Reference standard
  • CAS No.:
    1531-25-5
  • Description:
    Dextromethorphan impurity A
  • Formula:
    C₁₇H₂₃NO
  • Storage temperature:
    Ambient
  • Description:
    Dextromethorphan impurity A

Specifications

About this item

  • In Monograph 0020