Organic reference standards, Amiodarone, Ph.Eur., RS
Supplier: EDQM
Certificates
About this item
- Method: Monograph 0803
- Pharmaceutical industry applications
Specifications
- CAS No.:1951-26-4
- Formula:C₁₉H₁₆I₂O₃
- Storage temperature:Ambient
- Description:Amiodarone impurity D
Specifications
Frequently Bought Together