Organic reference standards, Acetylcysteine impurity, Ph.Eur., RS
Supplier: EDQM
Certificates
About this item
Specifications
- CAS No.:5545-17-5
- Formula:C₁₀H₁₆N₂O₆S₂
- Storage temperature:+5°C ± 3°C
- Description:Acetylcysteine impurity C CRS
Specifications
Frequently Bought Together