ContactUsLinkComponent
DNAzure® Blue Nucleic acid gel stain, 100X, Biotium
DNAzure® Blue Nucleic acid gel stain, 100X, Biotium
Catalog # 41020.
Supplier:  Biotium
CAS Number:  
DNAzure® Blue Nucleic acid gel stain, 100X, Biotium
Catalog # 41020.
Supplier:  Biotium
CAS Number:  
Specifications
  • Pk:
    10 mL
  • Description:
    DNAzure® 100X
  • Seq:
    02
Specifications
About this item
DNAzure® Blue Nucleic Acid Gel Stain is an ultrasensitive reagent for visible staining of dsDNA in agarose or polyacrylamide gels.
  • Deep blue bands visible by the naked eye following 5-30 min light exposure
  • Ultrasensitive detection, as little as ~1 ng DNA
  • Simplified DNA band excision, without the need for DNA-damaging UV light
  • Bands are stable for weeks after color development
  • Compatible with downstream applications such as sequencing and cloning
  • Expensive gel documentation systems not required for imaging
After incubation with DNAzure® Blue Nucleic Acid Gel Stain, the gel must be exposed to bright light to facilitate the development of dark blue DNA bands.