Dutasteride, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
3 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1229922.
- 1229931.
- 1200917.
- Description:
- Dutasteride
- Dutasteride Resolution Mixture
- Dihydrodutasteride
- Synonym(s):
- N-[2,5-Bis(trifluoromethyl)phenyl]-3-oxo-4-aza-5α-androst-1-ene-17β-carboxamide
- CAS No.:
- 164656-23-9
- 164656-22-8
- Formula:
- C₂₇H₃₀F₆N₂O₂
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1229922.DutasterideSpecifications:
More
Specifications
Cat. No.1229922.DescriptionDutasterideSynonym(s)CAS No.164656-23-9FormulaC₂₇H₃₀F₆N₂O₂ - 1229931.Dutasteride Resolution MixtureDutasteride Resolution MixtureUSP reference standardVial20 mgLog in for availability1 * 20 mg1 560.00 SFr.Specifications:
More
Specifications
Cat. No.1229931.DescriptionDutasteride Resolution MixtureSynonym(s)CAS No.Formula - 1200917.DihydrodutasterideSpecifications:
More
Specifications
Cat. No.1200917.DescriptionDihydrodutasterideSynonym(s)N-[2,5-Bis(trifluoromethyl)phenyl]-3-oxo-4-aza-5α-androst-1-ene-17β-carboxamideCAS No.164656-22-8Formula



