ContactUsLinkComponent
Aspirator bottles
Aspirator bottles
  BEHRQL9002607
 :  BEHR
Aspirator bottles
  BEHRQL9002607
 :  BEHR
  • Ext.Ø:
    210 mm
  • Capacity:
    10000 ml
  • Height:
    427 mm
  • Pk:
    1
Aspirator bottles, HDPE natural, narrow-neck storage bottles with scale.
  • UV protection for long life
  • Resistant to acids and chemicals
  • Special colour and printing possible
Storage vessel with drain made of food-safe polyethylene.