Order Entry
Switzerland
ContactUsLinkComponent
UV/Visible spectrophotometer, photoLab® 7600
UV/Visible spectrophotometer, photoLab® 7600
Catalog # 705-0922
Supplier:  WTW
undefined
UV/Visible spectrophotometer, photoLab® 7600
Catalog # 705-0922
Supplier:  WTW

Specifications

  • Model:
    7600
  • Range:
    190 - 1100 nm
  • Light source:
    Xenon flash
  • Description:
    UV/Vis spectrophometer
  • Scanning speed:
    700 - 2000 nm/min in 1, 2, 5, 10 nm steps of wavelength range
  • W×D×H:
    404×314×197 mm
  • Wavelength range:
    190 - 1100 nm
  • Wavelength accuracy:
    ±0,5 nm
  • Display:
    Backlit 7” colour display for extensive graphical evaluation
  • Interfaces:
    2× USB, Ethernet
  • Measuring modes:
    Absorbance, % transmission, concentration, spectrum, kinetics, OptRF
  • No. of methods:
    Over 200 pre-programmed methods, 1000 user-defined methods, 20 profiles for kinetics and spectra, comprehensive programming options
  • Optical design:
    Monochromator with grating and step motor / reference beam
  • Scan:
    700 - 2000 nm/min, Scans in 1, 2, 5, 10 nm steps of wavelength range
  • Spectral bandwidth:
    4 nm
  • Storage capacity:
    5000; 500 spectra (300 - 900 nm); 400 kinetics with 150 values
  • Weight:
    4,5 kg
  • Pk:
    1

Specifications

About this item

The pHotoLab® 7600 combines routine analysis with spectral analysis and pioneering procedure OptRF.

  • Pioneering measurement procedure 'OptRF', reagent-free for COD, nitrate and nitrite in the UV range
  • Brilliant colour screen operation guidance
  • Multi-step and multi-wavelength programming tools
  • photoLab® color+data management included
  • Routine analysis with barcode recognition and automatic measurement range adaption for all cuvette sizes

The important parameters COD, nitrate and nitrite of municipal waste water samples from the outlet can be measured simply and directly via the optical reagent-free measurement (OptRF) in the UV range. Pipette sample into cuvette, put them into the meter, read result. Handy way for retainer samples or a quick check without digestion.

Certifications: CIE-compliant and other colour measurement options with optional PC software package photoLab® color+Data spectral.