Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Glyceryl Monooleate, Reference Standards

Glyceryl Monooleate, Reference Standards

Supplier:   USP
Glyceryl Monooleate, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1295742.
  • 1295753.
  • Description:
  • Glyceryl Monooleate 90%
  • Glyceryl Monooleate 40%
  • CAS No.:
  • 25496-72-4
  • 25496-72-4
  • Formula:
  • C₂₁H₄₀O₄
  • C₂₁H₄₀O₄

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1295742.
    Glyceryl Monooleate 90%
    Glyceryl Monooleate 90%
    USP reference standard
    Vial
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 440.00 SFr.
    440.00 SFr.
    1 * 250 mg
       440.00 SFr.

    Specifications:
    More
    Specifications
    Cat. No.
    1295742.
    Description
    Glyceryl Monooleate 90%
    CAS No.
    25496-72-4
    Formula
    C₂₁H₄₀O₄
  • 1295753.
    Glyceryl Monooleate 40%
    Glyceryl Monooleate 40%
    USP reference standard
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 445.00 SFr.
    445.00 SFr.
    1 * 500 mg
       445.00 SFr.

    Specifications:
    More
    Specifications
    Cat. No.
    1295753.
    Description
    Glyceryl Monooleate 40%
    CAS No.
    25496-72-4
    Formula
    C₂₁H₄₀O₄
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.