Order Entry
Switzerland
ContactUsLinkComponent
Levetiracetam, Reference Standards
Levetiracetam, Reference Standards
Supplier:  USP
Certificates
Levetiracetam, Reference Standards
Supplier:  USP

Specifications

  • CAS No.:
    102767-28-2
  • Formula:
    C₈H₁₄N₂O₂
  • Description:
    Levetiracetam
  • Synonym(s):
    (S)-2-aminobutanamide hydrochloride

Specifications

Frequently Bought Together