Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Lovastatin, Reference Standards

Lovastatin, Reference Standards

Supplier:   USP
Lovastatin, Reference Standards
Supplier:  USP
Certificates

4 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Lovastatin Epimer Mixture: Epilovastatin and Lovastatin 1:1 Mixture

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1370600.
  • 1370611.
  • 1370655.
  • 1236833.
  • Description:
  • Lovastatin
  • Lovastatin Related Compound A
  • Lovastatin Impurity Identication Mixture
  • Lovastatin Epimer Mixture
  • Synonym(s):
  • 4,4a-Dihydrolovastatin
  • CAS No.:
  • 75330-75-5
  • 77517-29-4
  • Formula:
  • C₂₄H₃₆O₅
  • C₂₄H₃₈O₅

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1370600.
    Lovastatin
    Lovastatin
    USP reference standard
    Vial
    125 mg
    Log in for availability
    Log in for availability
    1 * 125 mg
    1 * 125 mg - SFr. 356.00
    SFr. 356.00
    1 * 125 mg
       SFr. 356.00

    Specifications:
    More
    Specifications
    Cat. No.
    1370600.
    Description
    Lovastatin
    Synonym(s)
    CAS No.
    75330-75-5
    Formula
    C₂₄H₃₆O₅
  • 1370611.
    Lovastatin Related Compound A
    Lovastatin Related Compound A
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - SFr. 1’420.00
    SFr. 1’420.00
    1 * 20 mg
       SFr. 1’420.00

    Specifications:
    More
    Specifications
    Cat. No.
    1370611.
    Description
    Lovastatin Related Compound A
    Synonym(s)
    4,4a-Dihydrolovastatin
    CAS No.
    77517-29-4
    Formula
    C₂₄H₃₈O₅
  • 1370655.
    Lovastatin Impurity Identication Mixture
    Lovastatin Impurity Identication Mixture
    USP reference standard
    Vial
    30 mg
    Log in for availability
    Log in for availability
    1 * 30 mg
    1 * 30 mg - SFr. 676.00
    SFr. 676.00
    1 * 30 mg
       SFr. 676.00

    Specifications:
    More
    Specifications
    Cat. No.
    1370655.
    Description
    Lovastatin Impurity Identication Mixture
    Synonym(s)
    CAS No.
    Formula
  • 1236833.
    Lovastatin Epimer Mixture
    Lovastatin Epimer Mixture
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - SFr. 889.00
    SFr. 889.00
    1 * 25 mg
       SFr. 889.00

    Specifications:
    More
    Specifications
    Cat. No.
    1236833.
    Description
    Lovastatin Epimer Mixture
    Synonym(s)
    CAS No.
    Formula