Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Alloys and Metal Ore Standards
Ferrotitanium alloy, reference standard, NCS
Ferrotitanium alloy, reference standard, NCS
Ferrotitanium alloy, reference standard, NCS
Catalog # PROONCSHC15601
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Expand image
Ferrotitanium alloy, reference standard, NCS
Catalog # PROONCSHC15601
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Specifications
Pk:
50 g
Description:
Ferrotitanium, NCS
Specifications
About this item
Product Details
13 analytes/elements measured
Product Details