Order Entry
Switzerland
ContactUsLinkComponent
Oil analysis standards, phosphorus in 20 blank oil, CONOSTAN®
Oil analysis standards, phosphorus in 20 blank oil, CONOSTAN®
Catalog # PROOCON150-100-155
CAS Number:  
Oil analysis standards, phosphorus in 20 blank oil, CONOSTAN®
Catalog # PROOCON150-100-155
CAS Number:  

Specifications

  • Concentration:
    1000 µg/g
  • Pk:
    50 g
  • Description:
    Phosphorus in 20 blank oil

Specifications

About this item