Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Organic reference standard, coumarin, Ph.Eur., RS
Organic reference standard, coumarin, Ph.Eur., RS
Organic reference standard, coumarin, Ph.Eur., RS
Catalog # EDQMY0000438
Supplier:
EDQM
CAS Number:
Expand image
Organic reference standard, coumarin, Ph.Eur., RS
Catalog # EDQMY0000438
Supplier:
EDQM
CAS Number:
Specifications
Pk:
110 mg
Standards:
Reference standard
CAS No.:
91-64-5
Description:
Coumarin
Formula:
C₉H₆O₂
Description:
Coumarin
Specifications
About this item
Product Details
Product Details