Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant NAD kinase (highly active) (from E. coli)
Human recombinant NAD kinase (highly active) (from E. coli)
Catalog # PRSI90-353
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant NAD kinase (highly active) (from E. coli)
Catalog # PRSI90-353
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His Tag
  • Storage conditions:
    Working aliquots are stable for up to 3 months when stored at –20 °C.
  • Enzyme name:
    NAD Kinase (highly active)
  • Enzyme activity:
    >1 U/mg protein. One unit is defined as the amount of enzyme that synthesizes 1 umol of NADP per min.
  • Purity:
    >95% (SDS-PAGE)
  • Sequence:
    1 - 446
  • Formulation:
    Liquid, Ammonium sulphate suspension.

Specifications

About this item

NAD kinase catalyses the transfer of a phosphate group from ATP to NAD+ to generate NADP+, which in its reduced form acts as an electron donor for biosynthetic reactions. NADP+ is an essential coenzyme in metabolism and provides reducing power to biosynthetic processes such as fatty acid biosynthesis.

Fusion-Tag: His Tag

This recombinant proteins is for research use only.