Human recombinant NAD kinase (catalytic domain, highly active) (from E. coli)
Catalog # PRSI90-352
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:His Tag
- Storage conditions:Working aliquots are stable for up to 3 months when stored at –20 °C.
- Enzyme name:NAD Kinase (catalytic domain, highly active)
- Enzyme activity:>2 U/mg protein. One unit is defined as the amount of enzyme that synthesizes 1 umol of NADP per min.
- Purity:>95% (SDS-PAGE)
- Sequence:64 - 446
- Formulation:Liquid, Ammonium sulphate suspension.
Specifications
About this item
NAD kinase catalyses the transfer of a phosphate group from ATP to NAD+ to generate NADP+, which in its reduced form acts as an electron donor for biosynthetic reactions. NADP+ is an essential coenzyme in metabolism and provides reducing power to biosynthetic processes such as fatty acid biosynthesis.
Fusion-Tag: His Tag
This recombinant proteins is for research use only.