Order Entry
Switzerland
ContactUsLinkComponent
Mouse recombinant carbonic anhydrase 12
Mouse recombinant carbonic anhydrase 12
Catalog # PRSI92-563
Supplier:  ProSci Inc.
CAS Number:  
Mouse recombinant carbonic anhydrase 12
Catalog # PRSI92-563
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Enzyme type:
    Recombinant
  • Species:
    Mouse
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
  • Enzyme name:
    Carbonic Anhydrase 12
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    32.4 kDa
  • Sequence:
    Ala25-Ser301
  • Formulation:
    Lyophilized from a 0,2 µm filtered solution of 20 mM Tris, 150 mM NaCl,pH 8,0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.

Specifications

About this item

Carbonic Anhydrase (CA) XII, also known as Car12 and CA12, is an extracellular enzyme involved in the regulation of the microenvironment acidity and tumour malignant phenotype, was originally identified as a protein overexpressed in some types of cancers. It has showed that CA XII is induced by hypoxia and oestrogen and expressed at high levels on various types of cancer. The enzyme is directly involved in tumour progression, and its inhibition has an anti-tumour effect. Apart from its role in carcinogenesis, the enzyme contributes to various other diseases like glaucoma and arteriosclerotic plaques, among others. CA XII is therefore regarded as promising target for specific therapies, and may be used as a novel prognostic marker in combination with histologic grade of the tumours.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.