Mouse recombinant carboxylesterase 3
Catalog # PRSI92-535
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Species:Mouse
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
- Enzyme name:Carboxylesterase 3
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:62.4 kDa
- Sequence:Tyr19-Glu561
- Formulation:Lyophilized from a 0,2 µm filtered solution of 20 mM Tris, 150 mM NaCl,pH 8,0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Specifications
About this item
Mouse Carboxylesterases 3 (CES3) is a member of five families of mammalian carboxylesterases that plays a role in catalysing hydrolytic and transesterification reactions with xenobiotics, anticancer pro-drugs and narcotics, and detoxifying organophosphates and insecticides. Mammalian carboxylesterases are enzymes with broad substrate specificities ranging from small molecule esters to longchain fatty acid esters. It is shown that CESs has key roles in the metabolism of a wide variety of clinical drugs, illicit narcotics and chemical nerve agents. CES3 is broadly expressed in liver, colon and brain.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.