Human recombinant Biliverdin reductase A (from E. coli)
Catalog # PRSI92-501
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
- Enzyme name:Biliverdin reductase A
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:33.8 kD
- Sequence:Glu6-Ser294
- Formulation:Lyophilized from a 0.2 µm filtered solution of 4mM HCl. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
Specifications
About this item
Human Biliverdin reductase A (BLVRA) is belonged to the Gfo/Idh/MocA family and Biliverdin reductase subfamily. BLVRA is an enzyme that in humans is encoded by the BLVRA gene. BLVRA plays an important role in reducing the gamma-methene bridge of the open tetrapyrrole, biliverdin IX alpha, to bilirubin with the concomitant oxidation of a NADH or NADPH cofactor. BLVRA acts on biliverdin by reducing its double-bond between the pyrrole rings into a single-bond. It accomplishes this using NADPH + H+ as an electron donor, forming bilirubin and NADP+ as products.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.