Specifications
- Pk:50 µG
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:N-6 His tag
- Storage conditions:Lyophilized protein should be stored at –20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4...7 °C for 2 - 7 days. Aliquots of reconstituted samples are stable at –20 °C for 3 months.
- Enzyme name:Sulfotransferase 1C4
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:14.1 kD
- Sequence:Met1-Lys102
- Formulation:Lyophilized from a 0.2 µm filtered solution of 4mM HCl. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
Specifications
About this item
Human Sulfotransferase (SULT1C4) is an enzyme that in humans is encoded by the SULT1C4 gene, belongs to the sulfotransferase 1 family. SULT1C4 is expressed at high levels in fetal lung and kidney and at low levels in fetal heart, adult kidney, ovary and spinal chord. Sulfotransferase utilises 3'-phospho-5'-adenylyl sulfate (PAPS) as sulfonate donor to catalyse the sulfate conjugation of drugs, xenobiotic compounds, hormones, and neurotransmitters. It shows activity towards p-nitrophenol and N-hydroxy-2-acetylamino-fluorene (N-OH-2AAF). SULT1C4 plays an important role incatalyzing the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.